siRNA Details
Gene Symbol: Smurf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109598.1 | NP_001103068.1 | 498 | |||
XM_006248882.1 | XP_006248944.1 | 494 | ||||
XM_008769002.1 | XP_008767224.1 | 494 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Smurf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109598.1 | NP_001103068.1 | 498 | |||
XM_006248882.1 | XP_006248944.1 | 494 | ||||
XM_008769002.1 | XP_008767224.1 | 494 |