siRNA Details
| Gene Symbol: Smurf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109598.1 | NP_001103068.1 | 941 | |||
| XM_006248882.1 | XP_006248944.1 | 1015 | ||||
| XM_008769002.1 | XP_008767224.1 | 937 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smurf1| Gene Symbol: Smurf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109598.1 | NP_001103068.1 | 941 | |||
| XM_006248882.1 | XP_006248944.1 | 1015 | ||||
| XM_008769002.1 | XP_008767224.1 | 937 | ||||