siRNA Details
| Gene Symbol: Tbc1d31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134842.1 | NP_001128314.1 | 1, 2 | 157 | ||
| XM_006241653.1 | XP_006241715.1 | 1, 2 | 87 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d31| Gene Symbol: Tbc1d31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134842.1 | NP_001128314.1 | 1, 2 | 157 | ||
| XM_006241653.1 | XP_006241715.1 | 1, 2 | 87 | |||