siRNA Details
| Gene Symbol: Rpgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773079.1 | XP_008771301.1 | 10 | 1523 | ||
| XM_008773081.1 | XP_008771303.1 | 10 | 1525 | |||
| XM_008773082.1 | XP_008771304.1 | 10 | 1524 | |||
| XM_008773083.1 | XP_008771305.1 | 10 | 1517 | |||
| GenBank mRNA | BC162028.1 | - | 10 | 1261 | 9100258 | |

Rpgr
Genomic Map