siRNA Details
| Gene Symbol: Fstl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053629.3 | NP_446081.1 | 2, 3 | 287 | ||
| GenBank mRNA | AB021295.1 | - | 2, 3 | 287 | ||
| AB071603.1 | - | 308 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fstl3| Gene Symbol: Fstl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053629.3 | NP_446081.1 | 2, 3 | 287 | ||
| GenBank mRNA | AB021295.1 | - | 2, 3 | 287 | ||
| AB071603.1 | - | 308 | ||||