siRNA Details
Gene Symbol: Alkbh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127450.1 | NP_001120922.1 | 7 | 912 | ||
GenBank mRNA | BC082048.1 | - | 7 | 878 | ||
BC098707.1 | - | 5 | 1423 |
Gene Symbol: LOC100912458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228800.2 | XP_006228862.1 | 7 | 950 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
