siRNA Details
| Gene Symbol: Eif4e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108808.1 | NP_001102278.1 | 5 | 958 | ||
| GenBank mRNA | BC158817.1 | - | 565 | 7374173 | ||
| FQ219916.1 | - | 638 | ||||
| FQ220431.1 | - | 681 | ||||
| FQ226325.1 | - | 650 | ||||
| FQ231706.1 | - | 650 | ||||
| FQ232529.1 | - | 650 | ||||
| FQ233479.1 | - | 586 | ||||

Eif4e2
Genomic Map