siRNA Details
| Gene Symbol: IQCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287501.1 | NP_001274430.1 | 20 | 2161 | ||
| NM_152558.4 | NP_689771.3 | 22 | 2255 | |||
| XM_017011902.1 | XP_016867391.1 | 22 | 2317 | |||
| XM_017011903.1 | XP_016867392.1 | 22 | 2195 | |||
| GenBank mRNA | AB028946.2 | - | 20 | 1958 | ||
| AL136792.1 | - | 21 | 1936 | |||
| BC043150.1 | - | 20 | 2081 | 5297871 | ||
| BC071858.1 | - | 22 | 2229 | 5478116 | ||
| DA246430.1 | - | 5 | 433 | |||
| KJ534872.1 | - | 22 | 2045 | |||
| KJ534873.1 | - | 6 | 329 | |||
| KJ535081.1 | - | 20 | 1951 | |||

IQCE
Genomic Map