siRNA Details
| Gene Symbol: LOC100233206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY539901.1 | - | 3905 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100233206| Gene Symbol: LOC100233206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY539901.1 | - | 3905 | |||