siRNA Details
| Gene Symbol: Cox17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053540.2 | NP_445992.1 | 1, 2 | 198 | ||
| GenBank mRNA | AB032178.1 | - | 1, 2 | 142 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cox17| Gene Symbol: Cox17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053540.2 | NP_445992.1 | 1, 2 | 198 | ||
| GenBank mRNA | AB032178.1 | - | 1, 2 | 142 | ||