siRNA Details
| Gene Symbol: Agbl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100981.1 | NP_001094451.1 | 4 | 473 | ||
| GenBank mRNA | AY387075.1 | - | 4 | 473 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agbl4| Gene Symbol: Agbl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100981.1 | NP_001094451.1 | 4 | 473 | ||
| GenBank mRNA | AY387075.1 | - | 4 | 473 | ||