siRNA Details
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107490.3 | 1, 2 | 67 | |||
| XM_008758938.1 | XP_008757160.1 | 1, 2 | 260 | |||
| XM_008758939.1 | XP_008757161.1 | 1, 2 | 261 | |||
| XM_008758940.1 | XP_008757162.1 | 1, 2 | 261 | |||

Cic
Genomic Map