siRNA Details
| Gene Symbol: Krr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108094.2 | NP_001101564.1 | 2, 3 | 397 | ||
| GenBank mRNA | BC128768.1 | - | 2, 3 | 397 | 7125536 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krr1| Gene Symbol: Krr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108094.2 | NP_001101564.1 | 2, 3 | 397 | ||
| GenBank mRNA | BC128768.1 | - | 2, 3 | 397 | 7125536 | |