siRNA Details
| Gene Symbol: Ddx49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115023.1 | NP_001108495.1 | 8 | 924 | ||
| XM_008771088.1 | XP_008769310.1 | 8 | 1037 | |||
| GenBank mRNA | BC158855.1 | - | 8 | 924 | 7930984 | |
| FQ216881.1 | - | 6 | 2222 | |||

Ddx49
Genomic Map