siRNA Details
| Gene Symbol: Zbtb7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127375.1 | NP_001120847.1 | 4 | 1044 | ||
| GenBank mRNA | BC166476.1 | - | 4 | 1044 | 9101314 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb7c| Gene Symbol: Zbtb7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127375.1 | NP_001120847.1 | 4 | 1044 | ||
| GenBank mRNA | BC166476.1 | - | 4 | 1044 | 9101314 | |