siRNA Details
| Gene Symbol: Nfe2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247236.2 | XP_006247298.1 | 5 | 2383 | ||
| XM_006247237.2 | XP_006247299.1 | 5 | 2375 | |||
| XM_006247238.2 | XP_006247300.1 | 4 | 1545 | |||
| XM_006247239.2 | XP_006247301.1 | 4 | 1298 | |||
| GenBank mRNA | BC169095.1 | - | 1777 | |||

Nfe2l1
Genomic Map