siRNA Details
| Gene Symbol: Mthfd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108462.2 | NP_001101932.2 | 19 | 2092 | ||
| GenBank mRNA | BC166462.1 | - | 19 | 2092 | 9030223 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mthfd1l| Gene Symbol: Mthfd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108462.2 | NP_001101932.2 | 19 | 2092 | ||
| GenBank mRNA | BC166462.1 | - | 19 | 2092 | 9030223 | |