siRNA Details
| Gene Symbol: Bptf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768403.1 | XP_008766625.1 | 30 | 8867 | ||
| GenBank mRNA | BC160926.1 | - | 4 | 732 | 8364269 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bptf| Gene Symbol: Bptf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768403.1 | XP_008766625.1 | 30 | 8867 | ||
| GenBank mRNA | BC160926.1 | - | 4 | 732 | 8364269 | |