siRNA Details
| Gene Symbol: Tmem179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109572.1 | NP_001103042.1 | 4 | 463 | ||
| XM_006231001.2 | XP_006231063.1 | 5 | 1491 | |||
| GenBank mRNA | FQ225083.1 | - | 5 | 598 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem179b| Gene Symbol: Tmem179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109572.1 | NP_001103042.1 | 4 | 463 | ||
| XM_006231001.2 | XP_006231063.1 | 5 | 1491 | |||
| GenBank mRNA | FQ225083.1 | - | 5 | 598 | ||