siRNA Details
| Gene Symbol: Cebpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287577.1 | NP_001274506.1 | 1 | 1118 | ||
| NM_001287578.1 | NP_001274507.1 | 1 | 1118 | |||
| NM_001287579.1 | NP_001274508.1 | 1 | 1118 | |||
| NM_012524.3 | NP_036656.1 | 1 | 1118 | |||

Cebpa
Genomic Map