siRNA Details
| Gene Symbol: Atp5g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017311.1 | NP_059007.1 | 3 | 144 | ||
| XM_006247188.2 | XP_006247250.1 | 3 | 187 | |||
| XM_006247189.1 | XP_006247251.1 | 3 | 292 | |||
| GenBank mRNA | D13123.1 | - | 3 | 144 | ||
| FQ209790.1 | - | 3 | 162 | |||
| FQ210883.1 | - | 2 | 108 | |||
| FQ216886.1 | - | 3 | 162 | |||
| FQ217997.1 | - | 3 | 158 | |||

Atp5g1
Genomic Map