siRNA Details
| Gene Symbol: CALML3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005185.3 | NP_005176.1 | 1 | 1292 | ||
| GenBank mRNA | BC031889.1 | - | 1 | 1200 | 4777524 | |
| DR421766.1 | - | 1 | 319 | |||
| M36707.1 | - | 1 | 2152 | |||
| M58026.1 | - | 1 | 1216 | |||

CALML3
Genomic Map