siRNA Details
| Gene Symbol: Bccip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108505.2 | NP_001101975.2 | 1, 2 | 197 | ||
| GenBank mRNA | FQ227185.1 | - | 1, 2 | 188 | ||
| FQ228402.1 | - | 1, 2 | 195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bccip| Gene Symbol: Bccip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108505.2 | NP_001101975.2 | 1, 2 | 197 | ||
| GenBank mRNA | FQ227185.1 | - | 1, 2 | 188 | ||
| FQ228402.1 | - | 1, 2 | 195 | |||