siRNA Details
| Gene Symbol: Pde11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127480.1 | NP_001120952.1 | 9 | 1052 | ||
| NM_001127481.2 | NP_001120953.1 | 7 | 656 | |||
| NM_080893.1 | NP_543169.1 | 7 | 1730 | |||
| GenBank mRNA | AB059360.1 | - | 7 | 658 | ||
| AB059361.1 | - | 9 | 1052 | |||
| AB059362.1 | - | 7 | 1730 | |||

Pde11a
Genomic Map