siRNA Details
| Gene Symbol: Acad10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134537.1 | NP_001128009.1 | 12 | 1875 | ||
| XM_006249386.2 | XP_006249448.1 | 18 | 2960 | |||
| GenBank mRNA | BC168650.1 | - | 18 | 2995 | 7121002 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acad10| Gene Symbol: Acad10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134537.1 | NP_001128009.1 | 12 | 1875 | ||
| XM_006249386.2 | XP_006249448.1 | 18 | 2960 | |||
| GenBank mRNA | BC168650.1 | - | 18 | 2995 | 7121002 | |