siRNA Details
| Gene Symbol: Osbpl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750319.3 | XP_003750367.1 | 22, 23 | 2868 | ||
| XM_006241324.2 | XP_006241386.1 | 21, 22 | 3864 | |||
| XM_006241325.1 | XP_006241387.1 | 20, 21 | 2435 | |||
| XM_006241326.2 | XP_006241388.1 | 20, 21 | 3827 | |||
| XM_008765365.1 | XP_008763587.1 | 22, 23 | 2587 | |||
| XM_008765367.1 | XP_008763589.1 | 21, 22 | 9292 | |||
| GenBank mRNA | BC168899.1 | - | 3, 4 | 739 | 7455687 | |

Osbpl8
Genomic Map