siRNA Details
Gene Symbol: Slc24a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108051.1 | NP_001101521.1 | 9 | 991 | ||
XM_006240471.2 | XP_006240533.1 | 10 | 4083 | |||
XM_006240472.2 | XP_006240534.1 | 10 | 4083 | |||
GenBank mRNA | BC168870.1 | - | 9 | 896 | 7104573 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172152, BC137945, AK044239, AY156046, BC171939.
Panther Classification:
Gene Ontology Categories:
