siRNA Details
| Gene Symbol: Olr1381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000523.1 | NP_001000523.1 | 1 | 897 | ||
| Gene Symbol: Olr1382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214831.1 | NP_999996.1 | 1 | 897 | ||

Olr1381
Genomic Map