siRNA Details
| Gene Symbol: Mdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030859.2 | NP_110486.1 | 2, 3 | 236 | ||
| XM_006234591.2 | XP_006234653.1 | 3, 4 | 401 | |||
| XM_006234592.2 | XP_006234654.1 | 3, 4 | 349 | |||
| XM_006234593.2 | XP_006234655.1 | 3, 4 | 1026 | |||
| GenBank mRNA | AB025023.1 | - | 2, 3 | 236 | ||
| AF315950.1 | - | 3, 4 | 315 | |||
| BC157805.1 | - | 3, 4 | 267 | 9026085 | ||

Mdk
Genomic Map