siRNA Details
| Gene Symbol: Glb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127529.2 | NP_001121001.1 | 15 | 1652 | ||
| XM_006245177.2 | XP_006245239.1 | 15 | 1810 | |||
| XM_006245179.2 | XP_006245241.1 | 15 | 1603 | |||
| XM_006245180.2 | XP_006245242.1 | 15 | 1615 | |||
| XM_006245182.2 | XP_006245244.1 | 15 | 1501 | |||
| XM_008767223.1 | XP_008765445.1 | 14 | 2708 | |||
| XM_008767224.1 | XP_008765446.1 | 15 | 2225 | |||
| XM_008767225.1 | XP_008765447.1 | 12 | 2438 | |||
| GenBank mRNA | BC161869.1 | - | 15 | 1652 | 7109779 | |

Glb1l
Genomic Map