siRNA Details
Gene Symbol: Vom2r41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099500.1 | NP_001092970.1 | 6 | 1649 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
