siRNA Details
| Gene Symbol: Rsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014193.2 | NP_001014215.2 | 4 | 932 | ||
| GenBank mRNA | BC085888.1 | - | 3 | 2254 | 7192402 | |
| FQ230600.1 | - | 4 | 917 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsrp1| Gene Symbol: Rsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014193.2 | NP_001014215.2 | 4 | 932 | ||
| GenBank mRNA | BC085888.1 | - | 3 | 2254 | 7192402 | |
| FQ230600.1 | - | 4 | 917 | |||