siRNA Details
| Gene Symbol: LOC100125362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103354.2 | NP_001096824.1 | 2 | 442 | ||
| XM_006243064.2 | XP_006243126.1 | 2 | 732 | |||
| XM_008766199.1 | XP_008764421.1 | 2 | 592 | |||
| GenBank mRNA | BC092621.1 | - | 1 | 288 | 7313228 | |

LOC100125362
Genomic Map