siRNA Details
| Gene Symbol: Tap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032056.3 | NP_114445.2 | 7, 8 | 1368 | ||
| GenBank mRNA | BC091140.1 | - | 6, 7 | 1301 | 7381526 | |
| KC222910.1 | - | 6, 7 | 1279 | |||
| KC222911.1 | - | 6, 7 | 1279 | |||
| X75306.1 | - | 7, 8 | 1315 | |||
| X75307.1 | - | 7, 8 | 1315 | |||
| Gene Symbol: LOC103689996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772777.1 | XP_008770999.1 | 7, 8 | 1420 | ||

Tap2
Genomic Map