siRNA Details
| Gene Symbol: H2afj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109610.2 | NP_001103080.1 | 1 | 420 | ||
| GenBank mRNA | BC157816.1 | - | 1 | 420 | 9034669 | |
| Gene Symbol: LOC103690002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763447.1 | XP_008761669.1 | 1 | 459 | ||

H2afj
Genomic Map