siRNA Details
Gene Symbol: Spry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106427.1 | NP_001099897.1 | 3 | 506 | ||
GenBank mRNA | BC169059.1 | - | 3 | 1969 | 7938818 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106427.1 | NP_001099897.1 | 3 | 506 | ||
GenBank mRNA | BC169059.1 | - | 3 | 1969 | 7938818 |