siRNA Details
| Gene Symbol: Mrpl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106633.1 | NP_001100103.1 | 3, 4 | 465 | ||
| XM_006237805.2 | XP_006237867.1 | 3, 4 | 490 | |||
| GenBank mRNA | FQ214529.1 | - | 3, 4 | 452 | ||
| FQ216110.1 | - | 3, 4 | 481 | |||
| FQ216137.1 | - | 3, 4 | 431 | |||
| FQ221067.1 | - | 3, 4 | 450 | |||

Mrpl15
Genomic Map