siRNA Details
| Gene Symbol: Eif2ak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105744.2 | NP_001099214.2 | 18 | 2841 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2ak4| Gene Symbol: Eif2ak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105744.2 | NP_001099214.2 | 18 | 2841 | ||