siRNA Details
| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107075.1 | NP_001100545.1 | 8 | 1381 | ||
| XM_006247890.2 | XP_006247952.1 | 9 | 1988 | |||
| XM_006247891.2 | XP_006247953.1 | 8 | 1778 | |||
| GenBank mRNA | BC168919.1 | - | 8 | 1456 | ||

Foxk2
Genomic Map