siRNA Details
| Gene Symbol: Cdc42ep4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107063.2 | NP_001100533.1 | 2 | 647 | ||
| XM_006247688.2 | XP_006247750.1 | 2 | 1450 | |||
| XM_006247689.2 | XP_006247751.1 | 2 | 626 | |||
| XM_006247690.2 | XP_006247752.1 | 2 | 2036 | |||
| GenBank mRNA | BC161986.1 | - | 2 | 647 | 9030824 | |

Cdc42ep4
Genomic Map