siRNA Details
| Gene Symbol: Rhbdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106684.1 | NP_001100154.1 | 5 | 446 | ||
| XM_006238802.2 | XP_006238864.1 | 7 | 1054 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhbdl2| Gene Symbol: Rhbdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106684.1 | NP_001100154.1 | 5 | 446 | ||
| XM_006238802.2 | XP_006238864.1 | 7 | 1054 | |||