siRNA Details
| Gene Symbol: C8g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106555.1 | NP_001100025.1 | 4 | 444 | ||
| XM_006233581.2 | XP_006233643.1 | 4 | 829 | |||
| GenBank mRNA | FQ210613.1 | - | 4 | 404 | ||
| FQ218819.1 | - | 4 | 404 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8g| Gene Symbol: C8g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106555.1 | NP_001100025.1 | 4 | 444 | ||
| XM_006233581.2 | XP_006233643.1 | 4 | 829 | |||
| GenBank mRNA | FQ210613.1 | - | 4 | 404 | ||
| FQ218819.1 | - | 4 | 404 | |||