siRNA Details
Gene Symbol: Urm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137562.2 | NP_001131034.2 | 3 | 159 | ||
GenBank mRNA | BC169094.1 | - | 3 | 149 | 7933748 | |
CB313752.1 | - | 3 | 162 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Urm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137562.2 | NP_001131034.2 | 3 | 159 | ||
GenBank mRNA | BC169094.1 | - | 3 | 149 | 7933748 | |
CB313752.1 | - | 3 | 162 |