siRNA Details
| Gene Symbol: FKBP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015258.1 | NP_056073.1 | 21 | 2307 | ||
| XM_006717018.2 | XP_006717081.1 | 21 | 2194 | |||
| XM_006717019.1 | XP_006717082.1 | 20 | 2155 | |||
| XM_017014552.1 | XP_016870041.1 | 20 | 2125 | |||
| GenBank mRNA | AB014574.1 | - | 21 | 2234 | ||
| AK301208.1 | - | 8 | 1061 | |||
| AK303074.1 | - | 20 | 2155 | |||
| BC077732.1 | - | 5 | 520 | |||

FKBP15
Genomic Map