siRNA Details
| Gene Symbol: Twist2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021691.2 | NP_067723.1 | 2 | 807 | ||
| GenBank mRNA | CB757313.1 | - | 1 | 230 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Twist2| Gene Symbol: Twist2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021691.2 | NP_067723.1 | 2 | 807 | ||
| GenBank mRNA | CB757313.1 | - | 1 | 230 | ||