siRNA Details
| Gene Symbol: Trim14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107934.1 | NP_001101404.1 | 3 | 623 | ||
| XM_006238060.2 | XP_006238122.1 | 4 | 762 | |||
| XM_006238061.2 | XP_006238123.1 | 4 | 667 | |||
| XM_006238062.2 | XP_006238124.1 | 4 | 775 | |||
| GenBank mRNA | BC166777.1 | - | 4 | 702 | 7934596 | |

Trim14
Genomic Map