siRNA Details
| Gene Symbol: Ubr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234339.1 | XP_006234401.1 | 1 | 586 | ||
| XM_008761950.1 | XP_008760172.1 | 1 | 587 | |||
| XM_008761951.1 | XP_008760173.1 | 1 | 587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubr3| Gene Symbol: Ubr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234339.1 | XP_006234401.1 | 1 | 586 | ||
| XM_008761950.1 | XP_008760172.1 | 1 | 587 | |||
| XM_008761951.1 | XP_008760173.1 | 1 | 587 | |||