siRNA Details
| Gene Symbol: Qsox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109898.1 | NP_001103368.1 | 5 | 566 | ||
| NM_053431.3 | NP_445883.1 | 5 | 566 | |||
| GenBank mRNA | AB044285.1 | - | 5 | 566 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Qsox1| Gene Symbol: Qsox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109898.1 | NP_001103368.1 | 5 | 566 | ||
| NM_053431.3 | NP_445883.1 | 5 | 566 | |||
| GenBank mRNA | AB044285.1 | - | 5 | 566 | ||