siRNA Details
| Gene Symbol: Gnl3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081958.1 | NP_001075427.1 | 7 | 558 | ||
| GenBank mRNA | CA512184.1 | - | 2 | 182 | ||
| FQ223839.1 | - | 196 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnl3l| Gene Symbol: Gnl3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081958.1 | NP_001075427.1 | 7 | 558 | ||
| GenBank mRNA | CA512184.1 | - | 2 | 182 | ||
| FQ223839.1 | - | 196 | ||||