siRNA Details
| Gene Symbol: Zfp575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107489.1 | NP_001100959.1 | 4 | 903 | ||
| XM_006228430.2 | XP_006228492.1 | 4 | 847 | |||
| XM_008758928.1 | XP_008757150.1 | 4 | 943 | |||
| XM_008758929.1 | XP_008757151.1 | 4 | 874 | |||

Zfp575
Genomic Map